Assay Design



Gene Transcripts

Gene Symbol
rwdd1
Entrez Gene ID
448293
Gene Name
RWD domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 69288313 - 69302610 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005815.1 NP_001005815.1 4 - 5 502 84
XM_012962878.2 XP_012818332.1 4 - 5 437 84
GenBank mRNA BC075365.1 - 4 - 5 502 84
CR761667.2 - 4 - 5 474 84

View Full Product Details