Assay Design



Gene Transcripts

Gene Symbol
atf1
Entrez Gene ID
448303
Gene Name
activating transcription factor 1
Gene Aliases
ews-atf1, fus/atf-1, treb36
Chromosome Location
Chr.2: 140003616 - 140014443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005828.2 NP_001005828.1 3 - 4 346 67
GenBank mRNA BC075378.1 - 3 - 4 348 67
CT030449.1 - 3 - 4 350 67
CU075764.1 - 3 - 4 354 67

View Full Product Details