Assay Design



Gene Transcripts

Gene Symbol
myoz1
Entrez Gene ID
448363
Gene Name
myozenin 1
Gene Aliases
cs-2, fatz, myoz
Chromosome Location
Chr.7: 52857763 - 52878939 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006716.1 NP_001006717.1 2 - 3 239 111
XM_012966388.2 XP_012821842.1 2 - 3 261 111
GenBank mRNA BC075445.1 - 2 - 3 239 111

View Full Product Details