Assay Design



Gene Transcripts

Gene Symbol
bag2
Entrez Gene ID
448388
Gene Name
BCL2 associated athanogene 2
Gene Aliases
-
Chromosome Location
Chr.5: 88949264 - 88957168 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006728.1 NP_001006729.1 2 - 3 354 85
GenBank mRNA BC075476.1 - 2 - 3 354 85
CR760041.2 - 2 - 3 256 85

View Full Product Details