Assay Design



Gene Transcripts

Gene Symbol
thap1
Entrez Gene ID
448407
Gene Name
THAP domain containing, apoptosis associated protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 100588784 - 100595063 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006737.1 NP_001006738.1 1 - 2 249 82
GenBank mRNA BC075499.1 - 1 - 2 249 82
CR762254.1 - 1 - 2 110 82

View Full Product Details