Assay Design



Gene Transcripts

Gene Symbol
slc19a3
Entrez Gene ID
448431
Gene Name
solute carrier family 19 member 3
Gene Aliases
thtr2
Chromosome Location
Chr.5: 125162300 - 125170982 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006754.1 NP_001006755.1 1 - 2 302 73
GenBank mRNA BC075531.1 - 1 - 2 302 73

View Full Product Details