Assay Design



Gene Transcripts

Gene Symbol
clic4
Entrez Gene ID
448566
Gene Name
chloride intracellular channel 4
Gene Aliases
xclic4
Chromosome Location
Chr.5: 142987114 - 143049081 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006830.1 NP_001006831.1 5 - 6 844 81
XM_012970309.1 XP_012825763.1 6 - 7 1098 81
GenBank mRNA BC076899.1 - 5 - 6 844 81

View Full Product Details