Assay Design



Gene Transcripts

Gene Symbol
rexo4
Entrez Gene ID
448635
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
xpmc2, xpmc2h
Chromosome Location
Chr.Un|NW_016683633: 21961 - 27036 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006867.1 NP_001006868.1 1 - 2 391 63
GenBank mRNA BC076977.1 - 1 - 2 391 63
CR855796.2 - 1 - 2 369 63

View Full Product Details