Assay Design



Gene Transcripts

Gene Symbol
amacr
Entrez Gene ID
493271
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
-
Chromosome Location
Chr.1: 186316815 - 186330655 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007885.2 NP_001007886.1 2 - 3 308 98
GenBank mRNA BC080155.1 - 2 - 3 309 98

View Full Product Details