Assay Design



Gene Transcripts

Gene Symbol
amy2b
Entrez Gene ID
493323
Gene Name
amylase, alpha 2B (pancreatic)
Gene Aliases
-
Chromosome Location
Chr.4: 99687135 - 99702939 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007947.1 NP_001007948.1 10 - 11 1484 76
GenBank mRNA BC080454.1 - 10 - 11 1484 76
CR761580.2 - 10 - 11 1503 76

View Full Product Details