Assay Design



Gene Transcripts

Gene Symbol
dact1
Entrez Gene ID
493325
Gene Name
dishevelled-binding antagonist of beta-catenin 1
Gene Aliases
Dapper, XDpr, dact1-a, dact1-b, dapper1, dpr, dpr1, frodo, hdpr1, thyex3
Chromosome Location
Chr.8: 83050407 - 83056044 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007949.2 NP_001007950.1 3 - 4 864 77
GenBank mRNA BC080457.1 - 3 - 4 868 77
CR761983.1 - 3 - 4 868 77

View Full Product Details