Assay Design



Gene Transcripts

Gene Symbol
keap1
Entrez Gene ID
493386
Gene Name
kelch like ECH associated protein 1
Gene Aliases
INrf2, klhl19
Chromosome Location
Chr.3: 127228691 - 127232789 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008023.1 NP_001008024.1 - 2022 68
GenBank mRNA BC080903.1 - - 2022 68
CR855517.2 - - 2016 68

View Full Product Details