Assay Design



Gene Transcripts

Gene Symbol
sox11
Entrez Gene ID
493415
Gene Name
SRY-box 11
Gene Aliases
XLS13, XLS13b, sox11-a, sox11-b, xSOX-11, xls13a, xsox11
Chromosome Location
Chr.5: 132100841 - 132102848 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008052.1 NP_001008053.1 1 - 1 1153 73
GenBank mRNA BC080939.1 - 1 - 1 1153 73
CT010559.2 - 1 - 1 1175 73

View Full Product Details