Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011077.1 | NP_001011077.1 | 5 - 6 | 957 | 79 |
XM_012960669.2 | XP_012816123.1 | 5 - 6 | 1447 | 79 | |
GenBank mRNA | BC084462.1 | - | 5 - 6 | 957 | 79 |
BC158260.1 | - | 5 - 6 | 960 | 79 |