Assay Design



Gene Transcripts

Gene Symbol
eif4a1
Entrez Gene ID
496556
Gene Name
eukaryotic translation initiation factor 4A1
Gene Aliases
ddx2a, eif-4a, eif4a
Chromosome Location
Chr.Un|NW_016684078: 27224 - 31701 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011139.1 NP_001011139.1 1 - 2 77 92
GenBank mRNA BC084468.1 - 1 - 2 77 92
CR855769.2 - 1 - 2 76 92

View Full Product Details