Assay Design



Gene Transcripts

Gene Symbol
rps15
Entrez Gene ID
496609
Gene Name
ribosomal protein S15
Gene Aliases
rig
Chromosome Location
Chr.1: 80519271 - 80523274 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011187.2 NP_001011187.1 2 - 3 111 61
GenBank mRNA BC086610.1 - 2 - 3 113 61

View Full Product Details