Assay Design



Gene Transcripts

Gene Symbol
reep5
Entrez Gene ID
496723
Gene Name
receptor accessory protein 5
Gene Aliases
reep6
Chromosome Location
Chr.1: 134697170 - 134722122 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011272.1 NP_001011272.1 1 - 2 209 100
GenBank mRNA BC087981.1 - 1 - 2 209 100
CR761929.2 - 1 - 2 179 100

View Full Product Details