Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011272.1 | NP_001011272.1 | 4 - 5 | 613 | 67 |
XM_018092847.1 | XP_017948336.1 | 4 - 5 | 437 | 67 | |
GenBank mRNA | BC087981.1 | - | 4 - 5 | 613 | 67 |
CR761929.2 | - | 4 - 5 | 583 | 67 |