Assay Design
Gene Transcripts
- Gene Symbol
- tmem47
- Entrez Gene ID
-
394494
- Gene Name
- transmembrane protein 47
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 103635192 - 103675503 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203568.1 |
NP_988899.1 |
2 - 3 |
393 |
82 |
GenBank mRNA |
BC059749.1 |
- |
2 - 3 |
393 |
82 |
View Full Product Details