Assay Design



Gene Transcripts

Gene Symbol
cxorf56
Entrez Gene ID
394969
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.8: 21033865 - 21101640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204012.2 NP_989343.1 4 - 5 454 95
XM_018096190.1 XP_017951679.1 4 - 5 510 95
GenBank mRNA BC064195.1 - 4 - 5 435 95
BC084906.1 - 4 - 5 454 95
CR760470.2 - 4 - 5 435 95

View Full Product Details