Assay Design



Gene Transcripts

Gene Symbol
mfsd14b
Entrez Gene ID
496797
Gene Name
major facilitator superfamily domain containing 14B
Gene Aliases
hiatl1
Chromosome Location
Chr.1: 86494668 - 86519320 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011334.1 NP_001011334.1 1 - 2 151 101
GenBank mRNA BC088481.1 - 1 - 2 151 101
CT027897.2 - 1 - 2 181 101

View Full Product Details