Assay Design



Gene Transcripts

Gene Symbol
timm22
Entrez Gene ID
496870
Gene Name
translocase of inner mitochondrial membrane 22 homolog
Gene Aliases
tex4, tim22
Chromosome Location
Chr.2: 26738185 - 26743272 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011397.1 NP_001011397.1 3 - 4 529 52
GenBank mRNA BC088600.1 - 3 - 4 529 52

View Full Product Details