Assay Design



Gene Transcripts

Gene Symbol
cfap300
Entrez Gene ID
496912
Gene Name
cilia and flagella associated protein 300
Gene Aliases
c11orf70
Chromosome Location
Chr.2: 159103065 - 159133016 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011429.1 NP_001011429.1 2 - 3 200 70
GenBank mRNA BC088061.1 - 2 - 3 200 70

Gene Symbol
LOC101734118
Entrez Gene ID
101734118
Gene Name
uncharacterized protein C11orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016685204: 2166 - 4690 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004920600.3 XP_004920657.3 2 - 3 266 70

View Full Product Details