Assay Design



Gene Transcripts

Gene Symbol
zcchc9
Entrez Gene ID
496956
Gene Name
zinc finger CCHC-type containing 9
Gene Aliases
-
Chromosome Location
Chr.1: 167894900 - 167899540 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011465.1 NP_001011465.1 3 - 4 576 83
GenBank mRNA BC088591.1 - 3 - 4 576 83

View Full Product Details