Assay Design



Gene Transcripts

Gene Symbol
cav1
Entrez Gene ID
548467
Gene Name
caveolin 1
Gene Aliases
Cav-1, bscl3, cav, cav1-a, cav1-b, caveolin-1, cgl3, mstp085, vip21
Chromosome Location
Chr.3: 58500799 - 58525112 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015750.1 NP_001015750.1 2 - 3 226 63
GenBank mRNA BC089686.1 - 2 - 3 226 63

View Full Product Details