Assay Design



Gene Transcripts

Gene Symbol
agtrap
Entrez Gene ID
548468
Gene Name
angiotensin 2 receptor associated protein
Gene Aliases
atrap
Chromosome Location
Chr.7: 54050063 - 54074323 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015751.1 NP_001015751.1 2 - 3 94 102
GenBank mRNA BC089687.1 - 2 - 3 94 102
CR761507.2 - 2 - 3 73 102

View Full Product Details