Assay Design



Gene Transcripts

Gene Symbol
tfpi2
Entrez Gene ID
548483
Gene Name
tissue factor pathway inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.6: 24332694 - 24343723 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015766.1 NP_001015766.1 4 - 5 714 65
GenBank mRNA BC089705.1 - 4 - 5 714 65

View Full Product Details