Assay Design



Gene Transcripts

Gene Symbol
cyp3a4
Entrez Gene ID
548503
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
-
Chromosome Location
Chr.5: 81217902 - 81239942 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015786.1 NP_001015786.1 5 - 6 468 86
GenBank mRNA BC089731.1 - 5 - 6 468 86

View Full Product Details