Assay Design



Gene Transcripts

Gene Symbol
ror2
Entrez Gene ID
548551
Gene Name
receptor tyrosine kinase like orphan receptor 2
Gene Aliases
Xror2, bdb, bdb1, ntrkr2
Chromosome Location
Chr.1: 126675486 - 126764149 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015834.1 NP_001015834.1 2 - 3 580 83
XM_018092983.1 XP_017948472.1 2 - 3 226 83
GenBank mRNA BC090120.1 - 2 - 3 580 83

View Full Product Details