Assay Design
Gene Transcripts
- Gene Symbol
- yrdc
- Entrez Gene ID
-
548554
- Gene Name
- yrdC N6-threonylcarbamoyltransferase domain containing
- Gene Aliases
-
drip3, irip, sua5
- Chromosome Location
-
Chr.2: 57893500 - 57903443 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC090125.1 |
- |
5 - 6 |
834 |
75 |
View Full Product Details