Assay Design



Gene Transcripts

Gene Symbol
haus2
Entrez Gene ID
548581
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
cep27
Chromosome Location
Chr.8: 85295509 - 85304066 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015864.1 NP_001015864.1 5 - 6 513 84
GenBank mRNA BC090378.1 - 5 - 6 513 84

View Full Product Details