Assay Design



Gene Transcripts

Gene Symbol
sar1b
Entrez Gene ID
548588
Gene Name
secretion associated, Ras related GTPase 1B
Gene Aliases
cmrd, gtbpb, sara2
Chromosome Location
Chr.3: 17174513 - 17198284 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015871.1 NP_001015871.1 2 - 3 219 92
GenBank mRNA BC090805.1 - 2 - 3 219 92

View Full Product Details