Assay Design



Gene Transcripts

Gene Symbol
lhx8
Entrez Gene ID
548653
Gene Name
LIM homeobox 8
Gene Aliases
lhx7, x-Lhx7
Chromosome Location
Chr.4: 70892016 - 70901886 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015899.1 NP_001015899.1 5 - 6 743 71
XM_012961035.2 XP_012816489.1 4 - 5 1996 71
GenBank mRNA BC118739.1 - 5 - 6 725 71
CR926447.1 - 5 - 6 743 71

View Full Product Details