Assay Design



Gene Transcripts

Gene Symbol
cdc37
Entrez Gene ID
548666
Gene Name
cell division cycle 37
Gene Aliases
-
Chromosome Location
Chr.3: 127114831 - 127124010 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015912.1 NP_001015912.1 6 - 7 780 66
GenBank mRNA BC135957.1 - 6 - 7 802 66
CR926424.1 - 6 - 7 780 66

View Full Product Details