Assay Design



Gene Transcripts

Gene Symbol
tbpl2
Entrez Gene ID
550063
Gene Name
TATA-box binding protein like 2
Gene Aliases
tbp, tbp2, trf3, xTBPL2
Chromosome Location
Chr.8: 84265327 - 84280443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017309.2 NP_001017309.1 6 - 7 939 98
GenBank mRNA CR855514.2 - 6 - 7 939 98

View Full Product Details