Assay Design
Gene Transcripts
- Gene Symbol
- anp32b
- Entrez Gene ID
-
594955
- Gene Name
- acidic nuclear phosphoprotein 32 family member B
- Gene Aliases
-
phapi2, ssp29
- Chromosome Location
-
Chr.1: 95394141 - 95423799 on Build UCB_Xtro_10.0
- Gene Symbol
- eml2
- Entrez Gene ID
-
100495318
- Gene Name
- echinoderm microtubule associated protein like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 75512229 - 75574964 on Build UCB_Xtro_10.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CT030067.1 |
- |
- |
1348 |
75 |
View Full Product Details