Assay Design



Gene Transcripts

Gene Symbol
aoc3
Entrez Gene ID
594972
Gene Name
amine oxidase, copper containing 3
Gene Aliases
aoc2
Chromosome Location
Chr.10: 11044582 - 11058524 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030413.1 NP_001025584.1 3 - 4 2078 95
GenBank mRNA BC091028.1 - 3 - 4 2078 95

View Full Product Details