Assay Design



Gene Transcripts

Gene Symbol
hibadh
Entrez Gene ID
594992
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.6: 32621952 - 32707613 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030433.1 NP_001025604.1 4 - 5 480 84
GenBank mRNA BC091056.1 - 4 - 5 480 84

View Full Product Details