Assay Design



Gene Transcripts

Gene Symbol
nup58
Entrez Gene ID
595038
Gene Name
nucleoporin 58kDa
Gene Aliases
nupl1, pro2463
Chromosome Location
Chr.2: 153352377 - 153368176 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030479.1 NP_001025650.1 3 - 4 468 75
GenBank mRNA BC091643.1 - 3 - 4 468 75
CR942452.2 - 3 - 4 475 75

View Full Product Details