Assay Design



Gene Transcripts

Gene Symbol
mrgbp
Entrez Gene ID
595073
Gene Name
MRG/MORF4L binding protein
Gene Aliases
c20orf20, eaf7, urcc4
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030510.1 NP_001025681.1 - 458 143
GenBank mRNA BC093470.1 - - 458 143

View Full Product Details