Assay Design



Gene Transcripts

Gene Symbol
trim8
Entrez Gene ID
613104
Gene Name
tripartite motif containing 8
Gene Aliases
gerp, rnf27
Chromosome Location
Chr.7: 20946258 - 21002487 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032341.1 NP_001027512.1 3 - 4 1275 90
XM_012966677.2 XP_012822131.1 3 - 4 1430 90
GenBank mRNA BC098511.1 - 3 - 4 1275 90

View Full Product Details