Assay Design



Gene Transcripts

Gene Symbol
entpd7
Entrez Gene ID
548672
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1, NTPDase7
Chromosome Location
Chr.7: 27137843 - 27156935 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015918.2 NP_001015918.1 3 - 4 335 71
XM_012966551.2 XP_012822005.1 3 - 4 372 71
GenBank mRNA BC170622.1 - 2 - 3 298 71
CR926412.2 - 3 - 4 335 71
DQ118412.1 - 1 - 2 183 71

View Full Product Details