Assay Design



Gene Transcripts

Gene Symbol
entpd7
Entrez Gene ID
548672
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1, NTPDase7
Chromosome Location
Chr.7: 27137843 - 27156935 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015918.2 NP_001015918.1 6 - 7 801 73
XM_012966551.2 XP_012822005.1 6 - 7 838 73
GenBank mRNA BC170622.1 - 5 - 6 764 73
CR926412.2 - 6 - 7 801 73
DQ118412.1 - 4 - 5 649 73

View Full Product Details