Assay Design



Gene Transcripts

Gene Symbol
arsd
Entrez Gene ID
548706
Gene Name
arylsulfatase D
Gene Aliases
-
Chromosome Location
Chr.2: 38722615 - 38734829 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015952.2 NP_001015952.1 6 - 7 1069 90
GenBank mRNA BC171182.1 - 5 - 6 918 90
BC171184.1 - 5 - 6 918 90
CR848473.2 - 6 - 7 1069 90

View Full Product Details