Assay Design



Gene Transcripts

Gene Symbol
vps25
Entrez Gene ID
548832
Gene Name
vacuolar protein sorting 25 homolog
Gene Aliases
derp9, eap20, fap20
Chromosome Location
Chr.10: 11422348 - 11432652 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016078.2 NP_001016078.1 1 - 2 57 66
GenBank mRNA BC121525.1 - 1 - 2 59 66
CR762222.2 - 1 - 2 57 66

View Full Product Details