Assay Design



Gene Transcripts

Gene Symbol
vps25
Entrez Gene ID
548832
Gene Name
vacuolar protein sorting 25 homolog
Gene Aliases
derp9, eap20, fap20
Chromosome Location
Chr.10: 11422348 - 11432652 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016078.2 NP_001016078.1 3 - 4 264 84
GenBank mRNA BC121525.1 - 3 - 4 266 84
CR762222.2 - 3 - 4 264 84

View Full Product Details