Assay Design



Gene Transcripts

Gene Symbol
mrpl1
Entrez Gene ID
548880
Gene Name
mitochondrial ribosomal protein L1
Gene Aliases
-
Chromosome Location
Chr.1: 91220950 - 91253388 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016126.2 NP_001016126.1 5 - 6 605 89
GenBank mRNA BC098087.1 - 5 - 6 580 89
BC158989.1 - 5 - 6 583 89
BC170718.1 - 5 - 6 605 89
BC170720.1 - 5 - 6 605 89
CR762063.2 - 5 - 6 605 89

View Full Product Details