Assay Design



Gene Transcripts

Gene Symbol
prelid1
Entrez Gene ID
548927
Gene Name
PRELI domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 36686817 - 36694862 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016173.2 NP_001016173.1 5 - 6 900 78
GenBank mRNA BC158502.1 - 5 - 6 721 78
CR761878.2 - 5 - 6 900 78

View Full Product Details