Assay Design



Gene Transcripts

Gene Symbol
adprhl2
Entrez Gene ID
548938
Gene Name
ADP-ribosylhydrolase like 2
Gene Aliases
arh3
Chromosome Location
Chr.2: 55919112 - 55934191 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016184.2 NP_001016184.1 5 - 6 752 72
GenBank mRNA CR761827.2 - 5 - 6 752 72

View Full Product Details