Assay Design



Gene Transcripts

Gene Symbol
asb6
Entrez Gene ID
549129
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
Chromosome Location
Chr.8: 4610478 - 4614765 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016375.2 NP_001016375.1 5 - 6 649 91
GenBank mRNA BC170694.1 - 4 - 5 560 91
BC170698.1 - 4 - 5 560 91
CR761252.2 - 5 - 6 649 91

View Full Product Details